Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
IL34
Entrez Gene ID
508292
Gene Name
interleukin 34
Gene Aliases
C18H16ORF77, IL-34
Chromosome Location
Chr.18: 1510307 - 1570900 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001100324.2 NP_001093794.1 6 - 7 1076 61
GenBank mRNA BC146187.1 - 6 - 7 1077 61

View Full Product Details