Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CUEDC1
Entrez Gene ID
534625
Gene Name
CUE domain containing 1
Gene Aliases
-
Chromosome Location
Chr.19: 8670137 - 8758964 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001099064.1 NP_001092534.1 6 - 7 1146 93
GenBank mRNA BC142531.1 - 6 - 7 1146 93

View Full Product Details