Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
VEZF1
Entrez Gene ID
539120
Gene Name
vascular endothelial zinc finger 1
Gene Aliases
-
Chromosome Location
Chr.19: 8774656 - 8790252 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001102227.2 NP_001095697.1 4 - 5 1038 141
GenBank mRNA BC149697.1 - 4 - 5 1039 141

View Full Product Details