Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SCRN1
Entrez Gene ID
534933
Gene Name
secernin 1
Gene Aliases
-
Chromosome Location
Chr.4: 66355082 - 66421251 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001110803.2 NP_001104273.2 5 - 6 1091 68

View Full Product Details