The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001114515.2 | NP_001107987.1 | 1 - 2 | 130 | 62 |
GenBank mRNA | BC103251.1 | - | 1 - 2 | 132 | 62 |