Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TKTL2
Entrez Gene ID
509186
Gene Name
transketolase like 2
Gene Aliases
-
Chromosome Location
Chr.6: 2165705 - 2171074 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001046025.1 NP_001039490.1 1 - 1 2452 139
GenBank mRNA BC111320.1 - 1 - 1 2452 139

View Full Product Details