Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
SYNDIG1
Entrez Gene ID
541217
Gene Name
synapse differentiation inducing 1
Gene Aliases
DSPC2, TMEM90B
Chromosome Location
Chr.13: 42413804 - 42517965 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001076263.1 NP_001069731.1 4 - 4 1753 133
GenBank mRNA BC123666.1 - 4 - 4 1753 133

View Full Product Details