Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ANGPTL6
Entrez Gene ID
521472
Gene Name
angiopoietin like 6
Gene Aliases
AGF, Angpt16
Chromosome Location
Chr.7: 14654381 - 14663188 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001128070.1 NP_001121542.1 2 - 3 784 57
GenBank mRNA EU599229.1 - 2 - 3 784 57
JN848788.1 - 2 - 3 751 57
JN848789.1 - 2 - 3 751 57

View Full Product Details