Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
UBE2A
Entrez Gene ID
282107
Gene Name
ubiquitin conjugating enzyme E2 A
Gene Aliases
BHR6A, HR6A
Chromosome Location
Chr.X: 3930592 - 3947887 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105325.2 NP_001098795.1 1 - 2 186 64
GenBank mRNA BC102100.1 - 1 - 2 187 64

View Full Product Details