Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
HSTN
Entrez Gene ID
100302500
Gene Name
histatherin
Gene Aliases
-
Chromosome Location
Chr.6: 85459411 - 85489801 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001163414.1 NP_001156886.1 1 - 2 139 127
GenBank mRNA GQ204113.1 - 1 - 2 139 127

View Full Product Details