Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PDZD4
Entrez Gene ID
538475
Gene Name
PDZ domain containing 4
Gene Aliases
-
Chromosome Location
Chr.X: 37018129 - 37045965 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205543.1 NP_001192472.1 2 - 3 297 64

View Full Product Details