Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ZKSCAN2
Entrez Gene ID
539463
Gene Name
zinc finger with KRAB and SCAN domains 2
Gene Aliases
-
Chromosome Location
Chr.25: 22823163 - 22836634 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205670.1 NP_001192599.1 6 - 7 1984 77

View Full Product Details