Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
VPS37D
Entrez Gene ID
539811
Gene Name
VPS37D subunit of ESCRT-I
Gene Aliases
-
Chromosome Location
Chr.25: 33581569 - 33585572 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205718.2 NP_001192647.2 1 - 2 254 105

View Full Product Details