Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MFRP
Entrez Gene ID
522979
Gene Name
membrane frizzled-related protein
Gene Aliases
-
Chromosome Location
Chr.15: 29922657 - 29928545 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205900.1 NP_001192829.1 1 - 2 49 80

View Full Product Details