Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NIM1K
Entrez Gene ID
530210
Gene Name
NIM1 serine/threonine protein kinase
Gene Aliases
NIM1
Chromosome Location
Chr.20: 31464678 - 31542909 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205975.1 NP_001192904.1 1 - 2 984 103

View Full Product Details