Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MTHFD2L
Entrez Gene ID
536269
Gene Name
methylenetetrahydrofolate dehydrogenase (NADP+ dependent) 2 like
Gene Aliases
-
Chromosome Location
Chr.6: 89092317 - 89244968 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001206095.3 NP_001193024.1 5 - 6 766 97

View Full Product Details