Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PLEKHM3
Entrez Gene ID
533312
Gene Name
pleckstrin homology domain containing M3
Gene Aliases
PLEKHM1L
Chromosome Location
Chr.2: 96095228 - 96318519 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001206217.1 NP_001193146.1 4 - 5 2145 75

View Full Product Details