Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MZF1
Entrez Gene ID
523865
Gene Name
myeloid zinc finger 1
Gene Aliases
-
Chromosome Location
Chr.18: 65786615 - 65796747 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001206304.3 NP_001193233.3 4 - 5 795 114

View Full Product Details