Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
IL27
Entrez Gene ID
614927
Gene Name
interleukin 27
Gene Aliases
-
Chromosome Location
Chr.25: 26061018 - 26066688 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001164653.1 NP_001158125.1 1 - 2 30 67

View Full Product Details