Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SLC8B1
Entrez Gene ID
508887
Gene Name
solute carrier family 8 member B1
Gene Aliases
SLC24A6
Chromosome Location
Chr.17: 61286699 - 61316746 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001177596.1 NP_001171067.1 6 - 7 999 57
GenBank mRNA BF889830.1 - 3 - 4 268 57

View Full Product Details