Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PATZ1
Entrez Gene ID
532416
Gene Name
POZ/BTB and AT hook containing zinc finger 1
Gene Aliases
-
Chromosome Location
Chr.17: 70125940 - 70143759 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191197.3 NP_001178126.1 4 - 5 1646 66

View Full Product Details