Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
PTAR1
Entrez Gene ID
784830
Gene Name
protein prenyltransferase alpha subunit repeat containing 1
Gene Aliases
-
Chromosome Location
Chr.8: 45843557 - 45894476 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001206857.1 NP_001193786.1 7 - 8 985 71

View Full Product Details