Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HIRIP3
Entrez Gene ID
539704
Gene Name
HIRA interacting protein 3
Gene Aliases
-
Chromosome Location
Chr.25: 26267918 - 26271518 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192296.1 NP_001179225.1 1 - 2 717 67

View Full Product Details