Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NLRC4
Entrez Gene ID
512480
Gene Name
NLR family CARD domain containing 4
Gene Aliases
-
Chromosome Location
Chr.11: 14815606 - 14865198 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192323.2 NP_001179252.2 2 - 3 278 87

View Full Product Details