Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LIMD1
Entrez Gene ID
617525
Gene Name
LIM domains containing 1
Gene Aliases
-
Chromosome Location
Chr.22: 53633801 - 53707789 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001193163.1 NP_001180092.1 1 - 2 1401 68

View Full Product Details