Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ANP32A
Entrez Gene ID
538427
Gene Name
acidic nuclear phosphoprotein 32 family member A
Gene Aliases
ANP32D, HPPCn, LANP, PHAPI
Chromosome Location
Chr.10: 15613398 - 15652308 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001195019.1 NP_001181948.1 3 - 4 467 113

View Full Product Details