Product Details
- SNP ID
-
rs79690990
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:241544683 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TAAAGGTGAGTTCACAATCAGAACT[A/G]CCAAGCATACGAGCTTCCTACTGCA
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
BOK-AS1
PubMed Links
Gene Details
- Gene
- BOK-AS1
- Gene Name
- BOK antisense RNA 1
There are no transcripts associated with this gene.
View Full Product Details