Product Details

SNP ID
rs77607082
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:186148432 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACCAACTTCTTTTCCATTCAGGTC[A/G]GAGAGAACCTCTGGGAAGCAGCATG
Phenotype
MIM: 601854
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DGKG PubMed Links

Gene Details

Gene
DGKG
Gene Name
diacylglycerol kinase gamma
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080744.1 4454 UTR 3 NP_001074213.1
NM_001080745.1 4454 UTR 3 NP_001074214.1
NM_001346.2 4454 UTR 3 NP_001337.2

View Full Product Details