Product Details

SNP ID
rs75622628
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:96121971 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATCTCGTCTTGATAACATCAGCAG[A/G]GGTCACTAAAGATGCTGCAGGCATA
Phenotype
MIM: 603859
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SLC25A13 PubMed Links

Gene Details

Gene
SLC25A13
Gene Name
solute carrier family 25 member 13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001160210.1 1695 Missense Mutation CCT,TCT P541S NP_001153682.1
NM_014251.2 1695 Missense Mutation CCT,TCT P540S NP_055066.1
XM_006715831.3 1695 Missense Mutation CCT,TCT P551S XP_006715894.1
XM_011515727.2 1695 Intron XP_011514029.1
XM_017011663.1 1695 Missense Mutation CCT,TCT P537S XP_016867152.1
XM_017011664.1 1695 Missense Mutation CCT,TCT P256S XP_016867153.1
XM_017011665.1 1695 Missense Mutation CCT,TCT P256S XP_016867154.1

View Full Product Details