Product Details

SNP ID
rs76686991
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:66015906 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAGAGTTTGCAATGAGTGTGCAGT[A/G]GACAGTGAGAGAACTGCCCTGTGCT
Phenotype
MIM: 613945
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DNAJC5B PubMed Links

Gene Details

Gene
DNAJC5B
Gene Name
DnaJ heat shock protein family (Hsp40) member C5 beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033105.4 Intron NP_149096.2
XM_011517618.2 Intron XP_011515920.1
XM_011517619.2 Intron XP_011515921.1
XM_011517620.2 Intron XP_011515922.1
XM_011517621.2 Intron XP_011515923.1

View Full Product Details