Product Details

SNP ID
rs77755202
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:56162566 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTATAATAAGTCTTAAAATAAAAGG[C/T]ATAACCTATAAGAAAGTTTCTGTAA
Phenotype
MIM: 603026
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PLAG1 PubMed Links

Gene Details

Gene
PLAG1
Gene Name
PLAG1 zinc finger
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001114634.1 5681 UTR 3 NP_001108106.1
NM_001114635.1 5681 UTR 3 NP_001108107.1
NM_002655.2 5681 UTR 3 NP_002646.2
XM_011517544.2 5681 UTR 3 XP_011515846.1
XM_017013576.1 5681 UTR 3 XP_016869065.1
XM_017013577.1 5681 UTR 3 XP_016869066.1

View Full Product Details