Product Details

SNP ID
rs74568212
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:56165871 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACATTCTATTGCTAAATTTTTTAAT[A/G]AGCCAGATGTCAAAAGGTAGAGCCA
Phenotype
MIM: 603026
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PLAG1 PubMed Links

Gene Details

Gene
PLAG1
Gene Name
PLAG1 zinc finger
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001114634.1 2376 UTR 3 NP_001108106.1
NM_001114635.1 2376 UTR 3 NP_001108107.1
NM_002655.2 2376 UTR 3 NP_002646.2
XM_011517544.2 2376 UTR 3 XP_011515846.1
XM_017013576.1 2376 UTR 3 XP_016869065.1
XM_017013577.1 2376 UTR 3 XP_016869066.1

View Full Product Details