Product Details

SNP ID
rs77384420
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:34255228 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCCCAGCCTCTCCTGGGTAGCTTT[C/T]TGGAGGTGATCTCATTTGTAAGCTG
Phenotype
MIM: 613747 MIM: 609787
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KIF24 PubMed Links

Gene Details

Gene
KIF24
Gene Name
kinesin family member 24
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_194313.2 Intron NP_919289.2
XM_005251451.2 Intron XP_005251508.1
XM_006716764.2 Intron XP_006716827.1
XM_011517861.1 Intron XP_011516163.1
XM_011517863.2 Intron XP_011516165.1
XM_017014697.1 Intron XP_016870186.1
XM_017014698.1 Intron XP_016870187.1
Gene
UBAP1
Gene Name
ubiquitin associated protein 1
There are no transcripts associated with this gene.

View Full Product Details