Product Details
- SNP ID
-
rs76060730
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:84670873 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACGTCCTGCAAATGCAGTGCCTCTC[A/G]GATCTGGTGCAGCGACCCTTCTCCT
- Phenotype
-
MIM: 600456
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
NTRK2
PubMed Links
Gene Details
- Gene
- NTRK2
- Gene Name
- neurotrophic receptor tyrosine kinase 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001007097.2 |
1037 |
Missense Mutation |
CAG,CGG |
Q42R |
NP_001007098.1 |
NM_001018064.2 |
1037 |
Missense Mutation |
CAG,CGG |
Q42R |
NP_001018074.1 |
NM_001018065.2 |
1037 |
Missense Mutation |
CAG,CGG |
Q42R |
NP_001018075.1 |
NM_001018066.2 |
1037 |
Missense Mutation |
CAG,CGG |
Q42R |
NP_001018076.1 |
NM_001291937.1 |
1037 |
Missense Mutation |
CAG,CGG |
Q42R |
NP_001278866.1 |
NM_006180.4 |
1037 |
Missense Mutation |
CAG,CGG |
Q42R |
NP_006171.2 |
XM_005252001.2 |
1037 |
Missense Mutation |
CAG,CGG |
Q42R |
XP_005252058.1 |
XM_005252003.2 |
1037 |
Missense Mutation |
CAG,CGG |
Q42R |
XP_005252060.1 |
XM_005252004.2 |
1037 |
Missense Mutation |
CAG,CGG |
Q42R |
XP_005252061.1 |
XM_005252006.3 |
1037 |
Missense Mutation |
CAG,CGG |
Q42R |
XP_005252063.1 |
XM_005252007.3 |
1037 |
Missense Mutation |
CAG,CGG |
Q42R |
XP_005252064.1 |
XM_011518718.2 |
1037 |
Missense Mutation |
CAG,CGG |
Q42R |
XP_011517020.1 |
XM_011518720.2 |
1037 |
Missense Mutation |
CAG,CGG |
Q42R |
XP_011517022.1 |
XM_017014751.1 |
1037 |
Missense Mutation |
CAG,CGG |
Q42R |
XP_016870240.1 |
XM_017014752.1 |
1037 |
Missense Mutation |
CAG,CGG |
Q42R |
XP_016870241.1 |
XM_017014753.1 |
1037 |
Missense Mutation |
CAG,CGG |
Q42R |
XP_016870242.1 |
XM_017014754.1 |
1037 |
Missense Mutation |
CAG,CGG |
Q42R |
XP_016870243.1 |
XM_017014755.1 |
1037 |
Missense Mutation |
CAG,CGG |
Q42R |
XP_016870244.1 |
XM_017014756.1 |
1037 |
Missense Mutation |
CAG,CGG |
Q42R |
XP_016870245.1 |
XM_017014757.1 |
1037 |
Missense Mutation |
CAG,CGG |
Q42R |
XP_016870246.1 |
XM_017014758.1 |
1037 |
Missense Mutation |
CAG,CGG |
Q42R |
XP_016870247.1 |
XM_017014759.1 |
1037 |
Missense Mutation |
CAG,CGG |
Q42R |
XP_016870248.1 |
XM_017014760.1 |
1037 |
Missense Mutation |
CAG,CGG |
Q42R |
XP_016870249.1 |
XM_017014761.1 |
1037 |
Missense Mutation |
CAG,CGG |
Q42R |
XP_016870250.1 |
View Full Product Details