Product Details

SNP ID
rs75666336
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:85948746 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGATACATGTGAAAGTTATATGTC[A/G]ATACCTTTATTTTATTTTATTTTTT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NAA35 PubMed Links

Gene Details

Gene
NAA35
Gene Name
N(alpha)-acetyltransferase 35, NatC auxiliary subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321881.1 Intron NP_001308810.1
NM_001321882.1 Intron NP_001308811.1
NM_024635.3 Intron NP_078911.3
XM_005252127.3 Intron XP_005252184.1
XM_011518903.2 Intron XP_011517205.1
XM_011518904.2 Intron XP_011517206.1
XM_017015012.1 Intron XP_016870501.1

View Full Product Details