Product Details

SNP ID
rs78546014
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:98732449 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGCACATTTGGAGGGCAGGTCCAT[C/T]GGCCACTCCTCACTTCTGTGGGCTC
Phenotype
MIM: 615370
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ANKS6 PubMed Links

Gene Details

Gene
ANKS6
Gene Name
ankyrin repeat and sterile alpha motif domain containing 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173551.4 1548 UTR 3 NP_775822.3
XM_005251793.4 1548 UTR 3 XP_005251850.1
XM_005251794.4 1548 UTR 3 XP_005251851.1
XM_006716998.3 1548 UTR 3 XP_006717061.1
XM_006716999.2 1548 UTR 3 XP_006717062.1
XM_011518357.2 1548 UTR 3 XP_011516659.1
XM_017014445.1 1548 UTR 3 XP_016869934.1
XM_017014446.1 1548 UTR 3 XP_016869935.1
XM_017014447.1 1548 UTR 3 XP_016869936.1

View Full Product Details