Product Details

SNP ID
rs75217962
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:104504293 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCGGCAAATTGGACATCTGTAAAA[C/G]TATTTTTCTTCCTGGGATTTTTTCA
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
OR13F1 PubMed Links

Gene Details

Gene
OR13F1
Gene Name
olfactory receptor family 13 subfamily F member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004485.1 31 Missense Mutation CTA,GTA L11V NP_001004485.1

View Full Product Details