Product Details

SNP ID
rs79836745
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:104504345 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACCCCAAAGTTCAGGTCATCATAT[C/T]TGCGGTGTGCTTGCTGATGTACCTG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR13F1 PubMed Links

Gene Details

Gene
OR13F1
Gene Name
olfactory receptor family 13 subfamily F member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004485.1 83 Missense Mutation TCT,TTT S28F NP_001004485.1

View Full Product Details