Product Details

SNP ID
rs78818776
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:124484330 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGTGGTGACCGCACCCCAAGGAGTC[A/G]TGTGCAATAGGCAGAAGTCAGGGAG
Phenotype
MIM: 184757
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ADGRD2 PubMed Links

Gene Details

Gene
ADGRD2
Gene Name
adhesion G protein-coupled receptor D2
There are no transcripts associated with this gene.

Gene
NR5A1
Gene Name
nuclear receptor subfamily 5 group A member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004959.4 Intron NP_004950.2
XM_005251871.3 Intron XP_005251928.1
XM_011518455.2 Intron XP_011516757.1

View Full Product Details