Product Details

SNP ID
rs75649875
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:127921222 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTTTCAGAGGAGGGCAGAGCTCAC[C/T]TGAGATCAGTAGTAGACCCCGTTCA
Phenotype
MIM: 612865 MIM: 606378
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PIP5KL1 PubMed Links

Gene Details

Gene
PIP5KL1
Gene Name
phosphatidylinositol-4-phosphate 5-kinase like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135219.1 2903 Intron NP_001128691.1
NM_173492.1 2903 Intron NP_775763.1
XM_017014279.1 2903 UTR 3 XP_016869768.1
Gene
ST6GALNAC4
Gene Name
ST6 N-acetylgalactosaminide alpha-2,6-sialyltransferase 4
There are no transcripts associated with this gene.

View Full Product Details