Product Details

SNP ID
rs75684290
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:77426105 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAGTGCCAGCTTCAGACCTGACAC[A/G]GCCACCTCAGTTGGTTATGGGCTTC
Phenotype
MIM: 604397 MIM: 605978
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GNA14 PubMed Links

Gene Details

Gene
GNA14
Gene Name
G protein subunit alpha 14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004297.3 Intron NP_004288.1
XM_011519224.1 Intron XP_011517526.1
XM_017015341.1 Intron XP_016870830.1
Gene
VPS13A
Gene Name
vacuolar protein sorting 13 homolog A
There are no transcripts associated with this gene.

View Full Product Details