Product Details

SNP ID
rs77963533
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:21133747 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAACCTATGTCTCAAAATAATAGCC[A/G]TGTAGGGTTTGGTTGGGTGATGTGG
Phenotype
MIM: 605491
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C10orf113 PubMed Links

Gene Details

Gene
C10orf113
Gene Name
chromosome 10 open reading frame 113
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001010896.2 Intron NP_001010896.2
NM_001177483.1 Intron NP_001170954.1
Gene
NEBL
Gene Name
nebulette
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001173484.1 Intron NP_001166955.1
NM_006393.2 Intron NP_006384.1
NM_213569.2 Intron NP_998734.1
XM_005252342.4 Intron XP_005252399.1
XM_005252343.4 Intron XP_005252400.1
XM_005252344.4 Intron XP_005252401.1
XM_011519291.2 Intron XP_011517593.1
XM_017015467.1 Intron XP_016870956.1
XM_017015468.1 Intron XP_016870957.1
XM_017015469.1 Intron XP_016870958.1

View Full Product Details