Product Details

SNP ID
rs74511331
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:48878716 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCGTCTGACCGAAGAACTCGGCTG[G/T]GATGAGATTATTGATAGAGAGTAGT
Phenotype
MIM: 605894
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
EID1 PubMed Links

Gene Details

Gene
EID1
Gene Name
EP300 interacting inhibitor of differentiation 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014335.2 624 Missense Mutation TGG,TGT W180C NP_055150.1
Gene
SHC4
Gene Name
SHC adaptor protein 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_203349.3 624 Intron NP_976224.3
XM_005254375.3 624 Intron XP_005254432.1

View Full Product Details