Product Details

SNP ID
rs76792095
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:3285647 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCCTTGGGTTGCCAAGAGTTAGGA[A/G]TGCCATTCTCATTATAATTTCTGTC
Phenotype
MIM: 604191
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF263 PubMed Links

Gene Details

Gene
ZNF263
Gene Name
zinc finger protein 263
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005741.4 Intron NP_005732.2
XM_006720831.1 Intron XP_006720894.1
XM_006720832.1 Intron XP_006720895.1
XM_006720833.1 Intron XP_006720896.1
XM_006720834.3 Intron XP_006720897.1
XM_011522344.1 Intron XP_011520646.1
XM_017022850.1 Intron XP_016878339.1

View Full Product Details