Product Details

SNP ID
rs78545913
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:4735010 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGGCGGCTCCGAGCCTCCCCTTCG[C/T]CTTCTGGATGCTCTGAGCTCCTCAA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ANKS3 PubMed Links

Gene Details

Gene
ANKS3
Gene Name
ankyrin repeat and sterile alpha motif domain containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001242929.1 113 Intron NP_001229858.1
NM_001308089.1 113 Intron NP_001295018.1
NM_001324129.1 113 Intron NP_001311058.1
NM_001324130.1 113 Intron NP_001311059.1
NM_133450.3 113 Intron NP_597707.1
XM_005255119.3 113 Intron XP_005255176.1
XM_005255121.3 113 Intron XP_005255178.1
XM_011522372.1 113 Intron XP_011520674.1
XM_011522373.1 113 Intron XP_011520675.1
XM_011522374.1 113 Intron XP_011520676.1
XM_011522378.1 113 Intron XP_011520680.1
XM_017022935.1 113 Intron XP_016878424.1
XM_017022936.1 113 Intron XP_016878425.1
Gene
C16orf71
Gene Name
chromosome 16 open reading frame 71
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_139170.2 113 Intron NP_631909.2
XM_005255144.3 113 Intron XP_005255201.1
XM_017022975.1 113 UTR 5 XP_016878464.1
XM_017022976.1 113 Intron XP_016878465.1

View Full Product Details