Product Details
- SNP ID
-
rs79197483
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:4502690 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CGTGGGATAAATGAGACTGTATCTC[A/G]AGCACCTGGGACGGGGCCAGATTCC
- Phenotype
-
MIM: 612584
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
SPNS2
PubMed Links
Gene Details
- Gene
- SPNS2
- Gene Name
- sphingolipid transporter 2
There are no transcripts associated with this gene.
View Full Product Details