Product Details

SNP ID
rs80268494
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:41828836 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTAGACACGATGTGGATGTTGGGG[G/T]GGAACACTTTCATCTGTCGGATAAT
Phenotype
MIM: 607063 MIM: 608778
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
FKBP10 PubMed Links

Gene Details

Gene
FKBP10
Gene Name
FK506 binding protein 10
There are no transcripts associated with this gene.

Gene
KLHL10
Gene Name
kelch like family member 10
There are no transcripts associated with this gene.

Gene
NT5C3B
Gene Name
5'-nucleotidase, cytosolic IIIB
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_052935.4 599 Missense Mutation CAC,CCC H174P NP_443167.4
XM_006721669.3 599 Missense Mutation CAC,CCC H131P XP_006721732.1
XM_006721670.3 599 Missense Mutation CAC,CCC H131P XP_006721733.1
XM_011524276.2 599 Missense Mutation CAC,CCC H174P XP_011522578.1
XM_011524277.2 599 Missense Mutation CAC,CCC H166P XP_011522579.1
XM_017024127.1 599 Intron XP_016879616.1

View Full Product Details