Product Details

SNP ID
rs79340044
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:41828839 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGACACGATGTGGATGTTGGGGTGG[A/G]ACACTTTCATCTGTCGGATAATTTC
Phenotype
MIM: 607063 MIM: 608778
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FKBP10 PubMed Links

Gene Details

Gene
FKBP10
Gene Name
FK506 binding protein 10
There are no transcripts associated with this gene.

Gene
KLHL10
Gene Name
kelch like family member 10
There are no transcripts associated with this gene.

Gene
NT5C3B
Gene Name
5'-nucleotidase, cytosolic IIIB
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_052935.4 596 Missense Mutation TCC,TTC S173F NP_443167.4
XM_006721669.3 596 Missense Mutation TCC,TTC S130F XP_006721732.1
XM_006721670.3 596 Missense Mutation TCC,TTC S130F XP_006721733.1
XM_011524276.2 596 Missense Mutation TCC,TTC S173F XP_011522578.1
XM_011524277.2 596 Missense Mutation TCC,TTC S165F XP_011522579.1
XM_017024127.1 596 Intron XP_016879616.1

View Full Product Details