Product Details

SNP ID
rs77319852
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:43875448 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGATGGCTGAGGGAGGGAACAGAG[A/G]AAGCGCACTGGGGCTGGGACTGAAT
Phenotype
MIM: 600723
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MPP2 PubMed Links

Gene Details

Gene
MPP2
Gene Name
membrane palmitoylated protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278370.1 4266 UTR 3 NP_001265299.1
NM_001278371.1 4266 UTR 3 NP_001265300.1
NM_001278372.1 4266 UTR 3 NP_001265301.1
NM_001278373.1 4266 UTR 3 NP_001265302.1
NM_001278374.1 4266 UTR 3 NP_001265303.1
NM_001278375.1 4266 UTR 3 NP_001265304.1
NM_001278376.2 4266 UTR 3 NP_001265305.2
NM_001278381.1 4266 UTR 3 NP_001265310.1
NM_005374.4 4266 UTR 3 NP_005365.4
XM_011524827.2 4266 UTR 3 XP_011523129.2

View Full Product Details