Product Details

SNP ID
rs76842131
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:36188986 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTACCACTCAGAAATAGAGGGGTAC[G/T]ATGAATGCCTACAGGTGCTGGGGAG
Phenotype
MIM: 613274
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
LOC101927809 PubMed Links

Gene Details

Gene
LOC101927809
Gene Name
uncharacterized LOC101927809
There are no transcripts associated with this gene.

Gene
MOCOS
Gene Name
molybdenum cofactor sulfurase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017947.2 Intron NP_060417.2

View Full Product Details